missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1-Methyladenine, 95%, Thermo Scientific Chemicals

Product Code. 10701671 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Packaging:
Glass bottle
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10701671 25 mg Glass bottle 25mg
10227073 100 mg Glass bottle 100mg
10398380 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10701671 Supplier Thermo Scientific Chemicals Supplier No. 201310250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5142-22-3 | C6H7N5 | 149.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5142-22-3
Molecular Formula C6H7N5
Molecular Weight (g/mol) 149.16
MDL Number MFCD00010532
InChI Key HPZMWTNATZPBIH-UHFFFAOYSA-N
Synonym 1-methyladenine, 1-methyl-1h-purin-6-amine, 1h-purin-6-amine, 1-methyl, adenine, 1-methyl, n1-methyladenine, 1-methyl-1h-purin-6-ylamine, 6h-purin-6-imine, 1,9-dihydro-1-methyl, 1-methyl-adenine, 1-methyl-9h-purin-6-imine, 1-methylhydropurine-6-ylamine
PubChem CID 78821
ChEBI CHEBI:18083
IUPAC Name 1-methylpurin-6-amine
SMILES CN1C=NC2=NC=NC2=C1N

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point >300.0°C
Color Beige or Yellow
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 25 mg
Beilstein 26, V,17, 136
Solubility Information Solubility in water: soluble
Formula Weight 149.15
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyladenine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.