missing translation for 'onlineSavingsMsg'
Learn More

1-Methyladenine, TRC

Product Code. 30012821
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012821 25 mg 25mg
30005076 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012821 Supplier Toronto Research Chemicals Supplier No. TRCM14393525MG
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5142-22-3
Molecular Formula C6 H7 N5
Molecular Weight (g/mol) 149.15
Synonym 1-Methyl-1H-purin-6-amine, 1-Methyl-6-aminopurine, 6-Amino-1-methylpurine
IUPAC Name 1-methylpurin-6-amine
SMILES Cn1cnc2ncnc2c1N

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C6H7N5/c1-11-3-10-6-4(5(11)7)8-2-9-6/h2-3H,7H2,1H3
Formula Weight 149.0701
Chemical Name or Material 1-Methyladenine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.