missing translation for 'onlineSavingsMsg'
Learn More

1-Methylbiguanide Hydrochloride, TRC

Product Code. 17980925
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17980925 25 mg 25mg
17993415 10 mg 10mg
17991825 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17980925 Supplier Toronto Research Chemicals Supplier No. TRCM26509525MG
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1674-62-0
Molecular Formula C3 H9 N5 . Cl H
Molecular Weight (g/mol) 151.6
Synonym 1-Methylbiguanide Hydrochloride, Metformin Hydrochloride Imp. E (EP) as Hydrochloride, Imidodicarbonimidic diamide, N-methyl-, hydrochloride (1:1) (ACI), Biguanide, 1-methyl-, hydrochloride (6CI, 7CI, 8CI), Imidodicarbonimidic diamide, N-methyl-, monohydrochloride (9CI), 1-Methylbiguanide Hydrochloride, N'-Methylbiguanide monohydrochloride
IUPAC Name 1-carbamimidoyl-3-methylguanidine;hydrochloride
SMILES Cl.CNC(=N)NC(=N)N

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C3H9N5.ClH/c1-7-3(6)8-2(4)5;/h1H3,(H6,4,5,6,7,8);1H
Formula Weight 151.0625
Percent Purity >95%
Chemical Name or Material 1-Methylbiguanide Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.