missing translation for 'onlineSavingsMsg'
Learn More

1-Methylimidazole, TRC

Product Code. 30041061
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041061 1 g 1g
30040370 10 g 10g
30010392 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041061 Supplier Toronto Research Chemicals Supplier No. TRCM3129801G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-47-7
Molecular Weight (g/mol) 82.1
SMILES Cn1ccnc1

Specifications

Molecular Formula C4H6N2
Quantity 1 g
Percent Purity >95%
Chemical Name or Material 1-Methylimidazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.