missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindole-2-boronic acid, 95%

Product Code. 15499797
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499797 1 g 1g
15409807 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499797 Supplier Thermo Scientific Alfa Aesar Supplier No. H53028.03
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 191162-40-0 | C9H10BNO2 | 174.99 g/mol

Reactant for Suzuki-Miyaura coupling. It ia an active Pharmaceutical ingredient hence used as pharmaceutical intermediates. Also used as rawv material in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant for Suzuki-Miyaura coupling. It ia an active Pharmaceutical ingredient hence used as pharmaceutical intermediates. Also used as rawv material in organic synthesis.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 191162-40-0
Molecular Formula C9H10BNO2
Molecular Weight (g/mol) 174.99
MDL Number MFCD01114668
InChI Key CBPBJUTWVXLSER-UHFFFAOYSA-N
Synonym 1-methyl-1h-indol-2-yl boronic acid, n-methylindole-2-boronic acid, 2-borono-1-methyl-1h-indole, 1-methylindol-2-yl boronic acid, 1-methyl-1h-indole-2-boronic acid, 1-methyl-1h-indol-2-yl-2-boronic acid, boronic acid,b-1-methyl-1h-indol-2-yl, boronic acid, 1-methyl-1h-indol-2-yl, acmc-209eur, n-methyl-indoleboronic acid
PubChem CID 22733820
SMILES CN1C(=CC2=CC=CC=C12)B(O)O

Specifications

Melting Point 108°C to 112°C
Quantity 1 g
IUPAC Name (1-methylindol-2-yl)boronic acid
Formula Weight 174.99
Percent Purity 95%
Chemical Name or Material 1-Methylindole-2-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.