missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindole-3-carbonitrile, 96%

Product Code. 15497087
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15497087 1 g 1g
15417097 250 mg 250mg
15407097 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15497087 Supplier Thermo Scientific Alfa Aesar Supplier No. H34157.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 24662-37-1 | C10H8N2 | 156.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24662-37-1
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.188
MDL Number MFCD00466602
InChI Key FBAXZPMXGBNBPE-UHFFFAOYSA-N
PubChem CID 2307681
IUPAC Name 1-methylindole-3-carbonitrile
SMILES CN1C=C(C2=CC=CC=C21)C#N

Specifications

Melting Point 55°C to 56°C
Quantity 1 g
Synonym 1-methyl-1h-indole-3-carbonitrile, 3-cyano-1-methylindole, 1h-indole-3-carbonitrile,1-methyl, 1h-indole-3-carbonitrile, 1-methyl, d06fsr, 3-cyano 1-methyl-1h-indole, fbaxzpmxgbnbpe-uhfffaoysa
Formula Weight 156.19
Percent Purity 96%
Chemical Name or Material 1-Methylindole-3-carbonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.