missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindole, 98%

Product Code. 11484334
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484334 10 g 10g
11494334 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484334 Supplier Thermo Scientific Alfa Aesar Supplier No. A12605.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 603-76-9 | C9H9N | 131.178 g/mol

1-Methylindole is useful for the determination of association constant for the electron-donor-acceptor complexes with 1-(2,4,6-trinitrophenyl) propan-2-one. It acts as a reactant for the preparation of polycyclic derivatives of indoles, bisindole derivatives and pharmaceutically active 2-oxo-1-pyrrolidine analogues. It is also used as a reactant for the preparation of Positron Emission Tomography (PET) imaging agents of protein kinase C (PKC) and glycogen synthase kinase-3 (GSK-3).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylindole is useful for the determination of association constant for the electron-donor-acceptor complexes with 1-(2,4,6-trinitrophenyl) propan-2-one. It acts as a reactant for the preparation of polycyclic derivatives of indoles, bisindole derivatives and pharmaceutically active 2-oxo-1-pyrrolidine analogues. It is also used as a reactant for the preparation of Positron Emission Tomography (PET) imaging agents of protein kinase C (PKC) and glycogen synthase kinase-3 (GSK-3).

Solubility
Immiscible with water.

Notes
Light sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-76-9
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00005800
InChI Key BLRHMMGNCXNXJL-UHFFFAOYSA-N
Synonym 1-methyl-1h-indole, n-methylindole, 1h-indole, 1-methyl, indole, 1-methyl, 1-methylindol, methylindole, unii-8h698roj5f, n-, chembl19912, n-methylindol
PubChem CID 11781
IUPAC Name 1-methylindole
SMILES CN1C=CC2=CC=CC=C21

Specifications

Density 1.048
Boiling Point 237°C to 239°C
Flash Point >110°C (230°F)
Odor Stench
Refractive Index 1.6065
Quantity 10 g
Beilstein 111026
Sensitivity Light sensitive
Solubility Information Immiscible with water.
Formula Weight 131.18
Percent Purity 98%
Chemical Name or Material 1-Methylindole
Show More Show Less
compliance-icons
Product Identifier
  • 1-Methylindole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1-Methylindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.