missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindoline-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10440574
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10440574 250 mg Amber Glass Bottle 250mg
10193074 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10440574 Supplier Thermo Scientific Maybridge Supplier No. CC58704CB
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 60082-02-2
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.204
MDL Number MFCD03208513
InChI Key OZQGVTARHLDSTI-UHFFFAOYSA-N
Synonym 1-methylindoline-5-carbaldehyde, 1-methyl-2,3-dihydro-1h-indole-5-carbaldehyde, 1-methylindoline-5-carboxaldehyde, 1-methyl-5-indolinecarbaldehyde, 1-methyl-5-formylindoline, 5-formyl-1-methylindoline, 1-methyl-5-indolinecarbaldehyde #, 2,3-dihydro-1-methyl-1h-indole-5-carboxaldehyde, 1h-indole-5-carboxaldehyde, 2,3-dihydro-1-methyl
PubChem CID 595181
IUPAC Name 1-methyl-2,3-dihydroindole-5-carbaldehyde
SMILES CN1CCC2=C1C=CC(=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 161.2
Percent Purity 97%
Chemical Name or Material 1-Methylindoline-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.