missing translation for 'onlineSavingsMsg'
Learn More

1-Methylisatin, 97%

Product Code. 11479773
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479773 1 g 1g
11489773 5 g 5g
11499773 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479773 Supplier Thermo Scientific Alfa Aesar Supplier No. A12942.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2058-74-4 | C9H7NO2 | 161.16 g/mol

1-Methylisatin is used as a reactant for stereoselective preparation of spirobicyclic and bis-spirotricyclic pyrazolidinones, for regioselective preparation of spirocyclic oxindole-butenolides, for synthesis of spiro-oxindoles, for preparation of unsymmetrical oxindoles, for stereoselective preparation of hydroxyloxindoles via Morita-Baylis-Hillman reaction, for preparation of pyridinecarboxylic acid [(oxo)indolylidene)hydrazide derivatives (Schiff base hydrazides) as antibacterial agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylisatin is used as a reactant for stereoselective preparation of spirobicyclic and bis-spirotricyclic pyrazolidinones, for regioselective preparation of spirocyclic oxindole-butenolides, for synthesis of spiro-oxindoles, for preparation of unsymmetrical oxindoles, for stereoselective preparation of hydroxyloxindoles via Morita-Baylis-Hillman reaction, for preparation of pyridinecarboxylic acid [(oxo)indolylidene)hydrazide derivatives (Schiff base hydrazides) as antibacterial agents.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2058-74-4
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00005812
InChI Key VCYBVWFTGAZHGH-UHFFFAOYSA-N
Synonym n-methylisatin, 1-methylisatin, 1-methyl-1h-indole-2,3-dione, 1-methylindoline-2,3-dione, 1h-indole-2,3-dione, 1-methyl, 1-methyl-indole-2,3-dione, 1 methylisatine, n-methylindol-2,3-dione, 1-methyl-2,3-dihydro-1h-indole-2,3-dione, 1-methyl-2,3-indolinedione
PubChem CID 16358
SMILES CN1C(=O)C(=O)C2=CC=CC=C12

Specifications

Melting Point 129°C to 133°C
Quantity 1 g
Beilstein 128280
Solubility Information Insoluble in water.
IUPAC Name 1-methylindole-2,3-dione
Formula Weight 161.16
Percent Purity 97%
Chemical Name or Material 1-Methylisatin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.