missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpiperidine-4-carboxylic acid hydrochloride, 96%

Product Code. 15458305
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458305 5 g 5g
15448305 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15458305 Supplier Thermo Scientific Alfa Aesar Supplier No. H33083.06
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 71985-80-3 | C7H14ClNO2 | 179.644 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71985-80-3
Molecular Formula C7H14ClNO2
Molecular Weight (g/mol) 179.644
MDL Number MFCD00190235
InChI Key NLUDEWJJEMHIIL-UHFFFAOYSA-N
Synonym 1-methylpiperidine-4-carboxylic acid hydrochloride, 1-methyl-piperidine-4-carboxylic acid hydrochloride, 1-methylpiperidine-4-carboxylic acid hcl, 4-carboxy-1-methyl-piperidine hcl, 1-methyl-4-piperidinecarboxylic acid hydrochloride, 1-methyl-piperidine-4-carboxylic acid hcl, 4-piperidinecarboxylic acid, 1-methyl-, hydrochloride, 1-methylpiperidine-4-carboxylic acid, chloride, 4-piperidinecarboxylicacid,1-methyl-,hydrochloride 1:1, pubchem8007
PubChem CID 2760043
IUPAC Name 1-methylpiperidine-4-carboxylic acid;hydrochloride
SMILES CN1CCC(CC1)C(=O)O.Cl

Specifications

Melting Point 225°C
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 179.65
Percent Purity 96%
Chemical Name or Material 1-Methylpiperidine-4-carboxylic acid hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.