missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpyrrole-2-carboxylic acid, 98%

Product Code. 11327635
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327635 5 g 5g
11317635 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327635 Supplier Thermo Scientific Alfa Aesar Supplier No. L05906.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6973-60-0 | C6H7NO2 | 125.127 g/mol

1-Methylpyrrole-2-carboxylic acid is used for thermochemical and computational studies of 1-Methyl-2-pyrrolecarboxylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylpyrrole-2-carboxylic acid is used for thermochemical and computational studies of 1-Methyl-2-pyrrolecarboxylic acid.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6973-60-0
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.127
MDL Number MFCD00003088
InChI Key ILAOVOOZLVGAJF-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrrole-2-carboxylic acid, 1-methyl-2-pyrrolecarboxylic acid, n-methylpyrrole-2-carboxylic acid, 1h-pyrrole-2-carboxylic acid, 1-methyl, 1h-pyrrolecarboxylic acid, methyl, 1-methyl-1h-pyrrole-2-carboxylicacid, pubchem9181, acmc-209oar, ksc353i0t, 2-carboxy-1-methyl-1h-pyrrole
PubChem CID 81453
IUPAC Name 1-methylpyrrole-2-carboxylic acid
SMILES CN1C=CC=C1C(=O)O

Specifications

Melting Point 135°C to 138°C
Quantity 5 g
Beilstein 115703
Formula Weight 125.13
Percent Purity 98%
Chemical Name or Material 1-Methylpyrrole-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.