missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpyrrolidine, 98%

Product Code. 11450258
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450258 100 g 100g
11460258 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450258 Supplier Thermo Scientific Alfa Aesar Supplier No. B23799.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 120-94-5 | C5H11N | 85.15 g/mol

1-Methylpyrrolidine is a methylated pyrollidine and is involved as an essential part of the structure of cefipime. It is also an active component of cigarette smoke.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylpyrrolidine is a methylated pyrollidine and is involved as an essential part of the structure of cefipime. It is also an active component of cigarette smoke.

Solubility
Miscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-94-5
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number MFCD00003173
InChI Key AVFZOVWCLRSYKC-UHFFFAOYSA-N
Synonym n-methylpyrrolidine, methylpyrrolidine, n-methyltetrahydropyrrole, pyrrolidine, 1-methyl, n-methyl pyrrolidine, 1-methyl-pyrrolidine, pyrrolidine, n-methyl, unii-06509tzu6c, methyl pyrrolidine
PubChem CID 8454
IUPAC Name 1-methylpyrrolidine
SMILES CN1CCCC1

Specifications

Melting Point °C to 90°C
Density 0.819
Boiling Point 80°C to 81°C
Flash Point -18°C (0°F)
Assay Percent Range 0.98
Refractive Index 1.4247
Quantity 100 g
UN Number UN3286
Solubility Information Miscible with water.
Formula Weight 85.15
Percent Purity 98%
Chemical Name or Material 1-Methylpyrrolidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.