missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpyrrolidine, TRC

Product Code. 30136612
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136612 10 g 10g
30012252 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136612 Supplier Toronto Research Chemicals Supplier No. TRCM32621010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-94-5
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
Synonym N-Methylpyrrolidine, N-Methyltetrahydropyrrole, NSC 65579;
SMILES CN1CCCC1

Specifications

Quantity 10 g
Chemical Name or Material 1-Methylpyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.