missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-3-methylimidazolium chloride, 96%

Product Code. 11349685
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11349685

Brand: Thermo Scientific Alfa Aesar L19749.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79917-90-1 | C8H15ClN2 | 174.67 g/mol

1-n-Butyl-3-methylimidazolium chloride is used as a solvent for organic reactions. It is used in anion exchange reactions. It acts as a catalyst, electrolyte for batteries and metal recovery. It is also used in chromatographic separations. Further, it is used as an ionic liquids for catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-n-Butyl-3-methylimidazolium chloride is used as a solvent for organic reactions. It is used in anion exchange reactions. It acts as a catalyst, electrolyte for batteries and metal recovery. It is also used in chromatographic separations. Further, it is used as an ionic liquids for catalysis.

Solubility
Soluble in acetone, acetonitrile, hot ethyl acetate, isopropyl alcohol, methylene chloride and methanol. Insoluble in water, hexane and toluene,

Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79917-90-1
Molecular Formula C8H15ClN2
Molecular Weight (g/mol) 174.67
MDL Number MFCD03095425
InChI Key FHDQNOXQSTVAIC-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium chloride, 3-butyl-1-methyl-1h-imidazol-3-ium chloride, 1-n-butyl-3-methylimidazolium chloride, bmimcl, bmim cl, c4mim chloride, dsstox_cid_11461, dsstox_rid_78878, dsstox_gsid_31461
PubChem CID 2734161
SMILES [Cl-].CCCCN1C=C[N+](C)=C1

Specifications

Melting Point ∼73°C
Quantity 50 g
Sensitivity Hygroscopic
Solubility Information Soluble in acetone,acetonitrile,hot ethyl acetate,isopropyl alcohol,methylene chloride and methanol. Insoluble in water,hexane and toluene,
IUPAC Name 1-butyl-3-methyl-1H-imidazol-3-ium chloride
Formula Weight 174.67
Percent Purity 96%
Chemical Name or Material 1-n-Butyl-3-methylimidazolium chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.