missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-3-methylimidazolium methyl sulfate, 99%

Product Code. 11382909
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382909 5 g 5g
11392909 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382909 Supplier Thermo Scientific Alfa Aesar Supplier No. H27754.06
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 401788-98-5 | C9H18N2O4S | 250.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 401788-98-5
Molecular Formula C9H18N2O4S
Molecular Weight (g/mol) 250.31
MDL Number MFCD03095437
InChI Key MEMNKNZDROKJHP-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium methyl sulfate, 1-butyl-3-methylimidazolium methylsulfate, bmimmeso4, 1-n-butyl-3-methylimidazolium methylsulfate, 1-butyl-3-methylimidazolium methyl sulfate 1-, acmc-20aowh, basionics™ lq 02, 1-butyl-3-methylimidazolium methylsul-fate, 1-butyl-3-methylimidazol-3-ium,methyl sulfate, 1-butyl-3-methyl-imidazol-3-ium; methyl sulfate
PubChem CID 12095228
SMILES COS([O-])(=O)=O.CCCCN1C=C[N+](C)=C1

Specifications

Density 1.21
Refractive Index 1.478
Quantity 5 g
UN Number UN3265
IUPAC Name 1-butyl-3-methyl-1H-imidazol-3-ium methyl sulfate
Formula Weight 250.32
Percent Purity 99%
Chemical Name or Material 1-n-Butyl-3-methylimidazolium methyl sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.