missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-3-methylimidazolium tetrafluoroborate, 98+%

Product Code. 11319645
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319645 50 g 50g
11309645 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319645 Supplier Thermo Scientific Alfa Aesar Supplier No. L19087.18
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 174501-65-6 | C8H15BF4N2 | 226.03 g/mol

1-n-Butyl-3-methylimidazolium tetrafluoroborate is an ionic liquid used for various reactions such as hydrogenations, asymmetric hydrogenations proceed in higher enantioselectivity than in homogeneous phase. It is also used in Suzuki cross-coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-n-Butyl-3-methylimidazolium tetrafluoroborate is an ionic liquid used for various reactions such as hydrogenations, asymmetric hydrogenations proceed in higher enantioselectivity than in homogeneous phase. It is also used in Suzuki cross-coupling.

Solubility
Miscible with acetone, acetonitrile, ethyl acetate, isopropyl alcohol and methylene chloride. Immiscible with hexane, toluene and water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174501-65-6
Molecular Formula C8H15BF4N2
Molecular Weight (g/mol) 226.03
MDL Number MFCD03095449
InChI Key LSBXQLQATZTAPE-UHFFFAOYSA-N
Synonym 1-butyl-3-methylimidazolium tetrafluoroborate, 1-n-butyl-3-methylimidazolium tetrafluoroborate, 3-butyl-1-methyl-1h-imidazol-3-ium tetrafluoroborate, unii-t2tvz2306t, bmimbf4, butylmethylimidazolium tetrafluoroborate, 1-butyl-3-methylimidazolium terafluoroborate, 1-methyl-3-butylimidazolium tetrafluoroborate, c4mim tetrafluoroborate, 1-butyl-3-methyl-1h-imidazol-3-ium tetrafluoroborate
PubChem CID 2734178
SMILES F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1

Specifications

Density 1.21
Flash Point None
Refractive Index 1.422
Quantity 50 g
Sensitivity Hygroscopic
Solubility Information Miscible with acetone,acetonitrile,ethyl acetate,isopropyl alcohol and methylene chloride. Immiscible with hexane,toluene and water.
IUPAC Name 1-butyl-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide
Formula Weight 226.03
Percent Purity ≥98%
Chemical Name or Material 1-n-Butyl-3-methylimidazolium tetrafluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.