missing translation for 'onlineSavingsMsg'
Learn More

1-N-Ethylgaramine Sulfate, TRC

Product Code. 30387341
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387341 50 mg -
30380450 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30387341 Supplier Toronto Research Chemicals Supplier No. TRCE91835050MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 864371-49-3
Molecular Formula C15H31N3O6 .xH2SO4
Molecular Weight (g/mol) 349.42
Synonym 2-Deoxy-6-O-[3-deoxy-4-C-methyl-3-(methylamino)-beta-L-arabinopyranosyl]-N1-ethyl-D-streptamine Sulfate Salt
IUPAC Name (2R,3R,4R,5R)-2-[(1R,2R,3R,4R,6R)-4-amino-6-(ethylamino)-2,3-dihydroxycyclohexyl]oxy-5-methyl-4-(methylamino)oxane-3,5-diol
SMILES CCN[C@@H]1C[C@@H](N)[C@@H](O)[C@@H](O)[C@@H]1O[C@H]2OC[C@](C)(O)[C@H](NC)[C@H]2O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H31N3O6/c1-4-18-8-5-7(16)9(19)10(20)12(8)24-14-11(21)13(17-3)15(2,22)6-23-14/h7-14,17-22H,4-6,16H2,1-3H3/t7-,8-,9-,10-,11-,12-,13-,14-,15+/m1/s1
Formula Weight 349.2213
Chemical Name or Material 1-N-Ethylgaramine Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.