missing translation for 'onlineSavingsMsg'
Learn More

1-n-Hexyl-3-methylimidazolium chloride, 98%

Product Code. 11446330
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446330 5 g 5g
11456330 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11446330 Supplier Thermo Scientific Alfa Aesar Supplier No. H27178.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 171058-17-6 | C10H19N2 | 167.28 g/mol

1-n-Hexyl-3-methylimidazolium chloride, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-n-Hexyl-3-methylimidazolium chloride, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 171058-17-6
Molecular Formula C10H19N2
Molecular Weight (g/mol) 167.28
MDL Number MFCD03093289
InChI Key RVEJOWGVUQQIIZ-UHFFFAOYSA-N
Synonym 1-hexyl-3-methylimidazolium chloride, hmimcl, 1-hexyl-3-methyl-1h-imidazol-3-ium chloride, 1-hexyl-3-methylimidazol-3-ium chloride, hmim cl, acmc-209e2c, dsstox_cid_27924, dsstox_rid_82677, dsstox_gsid_47948, ksc491c7f
PubChem CID 2734163
ChEBI CHEBI:61340
IUPAC Name 1-hexyl-3-methylimidazol-3-ium;chloride
SMILES CCCCCC[N+]1=CN(C)C=C1

Specifications

Refractive Index 1.5165
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 202.72
Percent Purity 98%
Chemical Name or Material 1-n-Hexyl-3-methylimidazolium chloride
compliance-icons
Product Identifier
  • 1-n-Hexyl-3-methylimidazolium chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1-Hexyl-3-methylimidazolium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.