missing translation for 'onlineSavingsMsg'
Learn More

1-n-Hexyl-3-methylimidazolium hexafluorophosphate, 99%

Product Code. 11372309
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372309 5 g 5g
11382309 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372309 Supplier Thermo Scientific Alfa Aesar Supplier No. H27272.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 304680-35-1 | C10H19F6N2P | 312.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304680-35-1
Molecular Formula C10H19F6N2P
Molecular Weight (g/mol) 312.24
MDL Number MFCD03093296
InChI Key YPWSSSRXUOQNMQ-UHFFFAOYSA-N
Synonym 1-hexyl-3-methylimidazolium hexafluorophosphate, 1-hexyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, 1-n-hexyl-3-methylimidazolium hexafluorophosphate, acmc-209hg2, dsstox_cid_27925, dsstox_rid_82678, dsstox_gsid_47949, ksc222c9l, 1-hexyl-3-methyl-imidazol-3-ium hexafluorophosphate, 1-hex-1-yl-3-methyl-1h-imidazol-3-ium hexafluorophosphate
PubChem CID 2734175
SMILES F[P-](F)(F)(F)(F)F.CCCCCCN1C=C[N+](C)=C1

Specifications

Refractive Index 1.419
Quantity 5 g
IUPAC Name 1-hexyl-3-methyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
Formula Weight 312.24
Percent Purity 99%
Chemical Name or Material 1-n-Hexyl-3-methylimidazolium hexafluorophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.