missing translation for 'onlineSavingsMsg'
Learn More

1-n-Propylcyclohexanol, 98%, Thermo Scientific Chemicals

Product Code. 11420007
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420007 5 g 5g
11430007 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420007 Supplier Thermo Scientific Alfa Aesar Supplier No. B20946.06
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5445-24-9
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00046364
InChI Key PYLPYOPJKOJRNP-UHFFFAOYSA-N
Synonym 1-propylcyclohexanol, 1-n-propylcyclohexanol, cyclohexanol, 1-propyl, 1-n-propyl cyclohexanol-1, propylcyclohexanol, 1-n-propyl-1-cyclohexanol, 1-n-propylcyclohexane-1-ol
PubChem CID 138504
IUPAC Name 1-propylcyclohexan-1-ol
SMILES CCCC1(O)CCCCC1

Specifications

Quantity 5 g
Formula Weight 142.24
Percent Purity 98%
Chemical Name or Material 1-n-Propylcyclohexanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.