missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthaleneacetic Acid, TRC

Product Code. 30009634
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009634 5 g 5g
30010154 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009634 Supplier Toronto Research Chemicals Supplier No. TRCN3776655G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-87-3
Molecular Formula C12 H10 O2
Molecular Weight (g/mol) 186.21
Synonym (Naphthalen-1-yl)acetic Acid, Naphazoline Nitrate Imp. B (EP), Naphazoline Hydrochloride Imp. B (EP), 1-Naphthylacetic Acid
IUPAC Name 2-naphthalen-1-ylacetic acid
SMILES OC(=O)Cc1cccc2ccccc12

Specifications

Quantity 5 g
InChI Formula InChI=1S/C12H10O2/c13-12(14)8-10-6-3-5-9-4-1-2-7-11(9)10/h1-7H,8H2,(H,13,14)
Formula Weight 186.0681
Percent Purity >95%
Chemical Name or Material 1-Naphthyl acetic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.