missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthalenemethanol, 98%

Product Code. 10701271 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 10701271

Brand: Thermo Scientific Acros 180110020

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4780-79-4 | C11H10O | 158.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4780-79-4
Molecular Formula C11H10O
Molecular Weight (g/mol) 158.20
MDL Number MFCD00004044
InChI Key PBLNHHSDYFYZNC-UHFFFAOYSA-N
Synonym 1-naphthalenemethanol, 1-naphthylmethanol, 1-hydroxymethylnaphthalene, naphthalene-1-methanol, 1-naphthalene methanol, 1-menaphthyl alcohol, 1-naphthylmethyl alcohol, naphthalen-1-yl-methanol, naphthalen-1-yl methanol, 1-naphthylenemethanol
PubChem CID 20908
ChEBI CHEBI:38137
IUPAC Name naphthalen-1-ylmethanol
SMILES OCC1=C2C=CC=CC2=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 59°C to 62°C
Color Yellow
Boiling Point 301°C (715.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C10H7CH2OH
Quantity 2 g
Beilstein 06, 667
Formula Weight 158.2
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 1-Naphthalenemethanol, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.