missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Naphthalenemethanol, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 1-Naphthalenemethanol |
| CAS | 4780-79-4 |
| Molecular Formula | C11H10O |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C11H10O/c12-8-10-6-3-5-9-4-1-2-7-11(9)10/h1-7,12H,8H2 |
| SMILES | OCc1cccc2ccccc12 |
| IUPAC Name | naphthalen-1-ylmethanol |
| Molecular Weight (g/mol) | 158.19 |
| Formula Weight | 158.0732 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction