missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 1-Naphthol

Product Code. 16402728
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16402728 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16402728 Supplier MedChemExpress Supplier No. HYY1309
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 1-naphthol is an excited state proton transfer (ESPT) fluorescent molecular probe.

Specifications

CAS 90-15-3
Molecular Weight (g/mol) 144.17 g/mol
Quantity 5 g
Shipping Condition Room temperature
Storage Requirements Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Solubility Ethanol : 100 mg/mL (693.63 mM; Need ultrasonic) ∣ H2O : 1 mg/mL (6.94 mM; Need ultrasonic)
Synonym Furro ER ∣ NSC 9586 ∣ Nako TRB
Target NO Synthase ∣ MMP ∣ Endogenous Metabolite
Product Type Natural Products
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.