missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthoyl Chloride, TRC

Product Code. 30464897
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464897 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464897 Supplier Toronto Research Chemicals Supplier No. TRCN49704025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Naphthoyl Chloride
CAS 879-18-5
Molecular Formula C11H7ClO
Quantity 25 g
Synonym 1-Naphthoyl Chloride, 1-(Chlorocarbonyl)naphthalene, 1-Naphthalenecarboxylic Acid Chloride, 1-Naphthoic Acid Chloride, 1-Naphthylcarbonyl Chloride, NSC 9841, α-Naphthoyl Chloride
InChI Formula InChI=1S/C11H7ClO/c12-11(13)10-7-3-5-8-4-1-2-6-9(8)10/h1-7H
SMILES ClC(=O)c1cccc2ccccc12
IUPAC Name naphthalene-1-carbonyl chloride
Molecular Weight (g/mol) 190.63
Formula Weight 190.0185
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.