missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl Acetate, TRC

Product Code. 30465629
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30465629 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30465629 Supplier Toronto Research Chemicals Supplier No. TRCN378675500MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Naphthyl Acetate
CAS 830-81-9
Molecular Formula C12 H10 O2
Quantity 500 mg
Synonym 1-Acetoxynaphthalene, 1-Naphthalen-1-yl Acetate, 1-Naphthyl Acetate, Acetic Acid 1-Naphthyl Ester, NSC 9846, Naphthalen-1-yl Acetate, α-Acetoxynaphthalene, α-Naphthol Acetate, α-Naphthyl Acetate
InChI Formula InChI=1S/C12H10O2/c1-9(13)14-12-8-4-6-10-5-2-3-7-11(10)12/h2-8H,1H3
SMILES CC(=O)Oc1cccc2ccccc12
IUPAC Name naphthalen-1-yl acetate
Molecular Weight (g/mol) 186.207
Formula Weight 186.068
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.