missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl isocyanate, 99%

Product Code. 10358953 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10358953 100 g 100g
10444311 5 g 5g
10454311 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10358953 Supplier Thermo Scientific Acros Supplier No. 415411000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 86-84-0 | C11H7NO | 169.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-84-0
InChI Key BDQNKCYCTYYMAA-UHFFFAOYSA-N
Synonym 1-naphthyl isocyanate, naphthalene, 1-isocyanato, alpha-naphthyl isocyanate, isocyanatonaphthalene, 1-naphthylisocyanate, 1-isocyanato-naphthalene, unii-5lh2p0691e, 1-isocyanatonapthalene, isocyanic acid 1-naphthyl ester, isocyanic acid, 1-naphthyl ester
PubChem CID 66589
IUPAC Name 1-isocyanatonaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2N=C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Density 1.18g/mL
Assay Percent Range 99%
Molecular Formula C11H7NO
Quantity 100 g
Merck Index 15, 6492
Molecular Weight (g/mol) 169.183
Formula Weight 169.18
Chemical Name or Material 1-Naphthyl isocyanate
compliance-icons
Product Identifier
  • 1-Naphthyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.