missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl phosphate monosodium salt monohydrate, 98+%

Product Code. 11423797
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423797 5 g 5g
11433797 25 g 25g
11443797 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423797 Supplier Thermo Scientific Alfa Aesar Supplier No. A18254.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 81012-89-7 | C10H7O4P | 222.14 g/mol

1-Naphthyl phosphate monosodium salt monohydrate acts as a substrate for determination of prostatic acid phosphatase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthyl phosphate monosodium salt monohydrate acts as a substrate for determination of prostatic acid phosphatase.

Solubility
Soluble in water.

Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81012-89-7
Molecular Formula C10H7O4P
Molecular Weight (g/mol) 222.14
MDL Number MFCD00150615
InChI Key YNXICDMQCQPQEW-UHFFFAOYSA-L
Synonym 1-naphthyl phosphate monosodium salt monohydrate, sodium naphthalen-1-yl hydrogenphosphate hydrate, alpha-naphthyl acid phosphate monosodium, sodium hydrate naphthalen-1-yl hydrogen phosphate, 1-naphthyl phosphate sodium salt monohydrate, c10h8po4.na.h2o, sodium 1-naphthyl phosphate monohydrate, 1-naphthyl phosphate, monosodium salt monohydrate, naphthalen-1-yloxy sodiooxy phosphinic acid hydrate, 1-naphthyl phosphate monosodium salt monohydrate hplc
PubChem CID 45055387
IUPAC Name naphthalen-1-yl dihydrogen phosphate;sodium;hydrate
SMILES [O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 186°C to 191°C
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 264.16 (246.14 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material 1-Naphthyl phosphate monosodium salt monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.