missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl phosphate, monosodium salt monohydrate, 98+%

Product Code. 15169253 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15169253 100 g Glass bottle 100g
10434345 1 g Glass bottle 1g
10124760 5 g Glass bottle 5g
10752685 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15169253 Supplier Thermo Scientific Acros Supplier No. 226891000
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 81012-89-7 | C10H7O4P | 222.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81012-89-7
Molecular Formula C10H7O4P
Molecular Weight (g/mol) 222.14
MDL Number MFCD00150615
InChI Key YNXICDMQCQPQEW-UHFFFAOYSA-L
Synonym 1-naphthyl phosphate monosodium salt monohydrate, sodium naphthalen-1-yl hydrogenphosphate hydrate, alpha-naphthyl acid phosphate monosodium, sodium hydrate naphthalen-1-yl hydrogen phosphate, 1-naphthyl phosphate sodium salt monohydrate, c10h8po4.na.h2o, sodium 1-naphthyl phosphate monohydrate, 1-naphthyl phosphate, monosodium salt monohydrate, naphthalen-1-yloxy sodiooxy phosphinic acid hydrate, 1-naphthyl phosphate monosodium salt monohydrate hplc
PubChem CID 45055387
SMILES [O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 189.0°C to 191.0°C
Color White
pH 3.5 to 4.5 (5% aq. soln.)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula C10H7OP(O)(OH)ONa·H2O
Quantity 100 g
Solubility Information (5% in water) Clear colorless to light yellow
IUPAC Name naphthalen-1-yl phosphate
Formula Weight 264.15
Percent Purity 98+%
Physical Form Fine Powder
Chemical Name or Material 1-Naphthyl phosphate, monosodium salt monohydrate, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Naphthyl phosphate, monosodium salt, monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.