missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthylacetyl chloride, 95%

Product Code. 11395369
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11395369 1 g 1g
11305379 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11395369 Supplier Thermo Scientific Alfa Aesar Supplier No. H50386.03
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 5121-00-6 | C12H9ClO | 204.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5121-00-6
Molecular Formula C12H9ClO
Molecular Weight (g/mol) 204.65
MDL Number MFCD00175852
InChI Key DSVAZLXLRDXHKO-UHFFFAOYSA-N
Synonym 2-1-naphthyl ethanoyl chloride, 2-naphthalen-1-yl acetyl chloride, 1-naphthylacetyl chloride, naphth-1-ylacetyl chloride, 1-naphthaleneacetyl chloride, naphth-1-yl acetyl chloride, napthylacetyl chloride, acmc-20aork, 1-naphthylacetylchloride, 1-naphthaleneacetylchloride
PubChem CID 2795386
IUPAC Name 2-(naphthalen-1-yl)acetyl chloride
SMILES ClC(=O)CC1=CC=CC2=CC=CC=C12

Specifications

Density 1.23
Boiling Point 130°C (0.6 mmHg)
Odor Pungent
Refractive Index 1.621
Quantity 1 g
UN Number UN3265
Formula Weight 204.66
Percent Purity 95%
Chemical Name or Material 1-Naphthylacetyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.