missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Nitronaphthalene-d7, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 1-Nitronaphthalene-d7 |
| CAS | 80789-77-1 |
| Molecular Formula | C10 D7 N O2 |
| Quantity | 0.05 g |
| Synonym | Naphthalene-1,2,3,4,5,6,7-d7, 8-nitro- (9 CI), 1-Nitronaphthalene-d7, 1-Nitronaphthalene-2,3,4,5,6,7,8-d7, 8-Nitronaphthalene-1,2,3,4,5,6,7-d7, 1-Nitronaphthalene-D7 |
| InChI Formula | InChI=1 S/C10H7NO2/c12-11(13)10-7-3-5-8-4-1-2-6-9(8)10/h1-7 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c2c(c([2 H])c([2 H])c([2 H])c2c1[2 H])[N+](=O)[O-] |
| IUPAC Name | 1,2,3,4,5,6,7-heptadeuterio-8-nitronaphthalene |
| Molecular Weight (g/mol) | 180.211 |
| Formula Weight | 180.092 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction