missing translation for 'onlineSavingsMsg'
Learn More

1-Nitropropane, 98%

Product Code. 11494150
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494150 100 g 100g
11414004 250 g 250g
11404160 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494150 Supplier Thermo Scientific Alfa Aesar Supplier No. A11975.22
 Restricted Item
In Stock
Add to basket
Læg i indkøbskurv
Request Bulk or Custom Format
Denne vare kan ikke returneres. Se returpolicy

CAS: 108-03-2 | C3H7NO2 | 89.094 g/mol

The solvent of 1-Nitropropane is mainly used coatings, inks and separation processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The solvent of 1-Nitropropane is mainly used coatings, inks and separation processes.

Solubility
Soluble in water. (1.4 g/L)

Notes
Store in cool place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 108-03-2
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.094
MDL Number MFCD00007407
InChI Key JSZOAYXJRCEYSX-UHFFFAOYSA-N
Synonym propane, 1-nitro, nitropropane, propane, nitro, ccris 1329, dsstox_cid_980, dsstox_rid_75902, dsstox_gsid_20980, nitro-propane, n-nitropropane, 3-nitropropane
PubChem CID 7903
ChEBI CHEBI:76261
IUPAC Name 1-nitropropane
SMILES CCC[N+](=O)[O-]

Tekniske data

Melting Point -108°C
Boiling Point 131°C to 132°C
Flash Point 34°C (93°F)
Odor Unpleasant
Linear Formula CH3CH2CH2NO2
Quantity 100 g
UN Number UN2608
Beilstein 506236
Merck Index 14,6627
Formula Weight 89.09
Percent Purity 98%
Chemical Name or Material 1-Nitropropane
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.