missing translation for 'onlineSavingsMsg'
Learn More

1-Nitropropane, 98%, Thermo Scientific Chemicals

Product Code. 10665562 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass bottle
Unit Size:
500g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10665562 500 g Glass bottle 500g
10396720 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10665562 Supplier Thermo Scientific Acros Supplier No. 128905000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-03-2
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00007407
InChI Key JSZOAYXJRCEYSX-UHFFFAOYSA-N
Synonym propane, 1-nitro, nitropropane, propane, nitro, ccris 1329, dsstox_cid_980, dsstox_rid_75902, dsstox_gsid_20980, nitro-propane, n-nitropropane, 3-nitropropane
PubChem CID 7903
ChEBI CHEBI:76261
IUPAC Name 1-nitropropane
SMILES CCC[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -108°C
Density 0.9900g/mL
Boiling Point 132°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2NO2
Refractive Index 1.4005 to 1.4025
Quantity 500 g
Beilstein 01, 115
Fieser 01,745
Merck Index 15, 6714
Specific Gravity 0.99
Solubility Information Solubility in water: 1.4mL/100mL (20°C). Other solubilities: miscible with many organic solvents
Viscosity 0.844 mPa.s (20°C)
Formula Weight 89.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Nitropropane, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.