missing translation for 'onlineSavingsMsg'
Learn More

1-Nitrosopyrrolidin-2-one (200 μg/mL in Methanol), TRC

Product Code. 30478259
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478259 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478259 Supplier Toronto Research Chemicals Supplier No. TRCKIT85151X1ML
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 54634-49-0
Molecular Formula C4H6N2O2
Quantity 1 mL
Synonym 1-Nitroso-2-pyrrolidinone, 1-Nitroso-2-pyrrolidone, N-Nitroso-2-pyrrolidinone
InChI Formula InChI=1S/C4H6N2O2/c7-4-2-1-3-6(4)5-8/h1-3H2
SMILES O=C1CCCN1N=O
IUPAC Name 1-nitrosopyrrolidin-2-one
Molecular Weight (g/mol) 114.1
Formula Weight 114.0429
Concentration or Composition (by Analyte or Components) 1-Nitrosopyrrolidin-2-one 200 μg/mL
Chemical Name or Material 1-Nitrosopyrrolidin-2-one
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.