missing translation for 'onlineSavingsMsg'
Learn More

1-Nonanethiol, 98%, Thermo Scientific™

Product Code. 11393116
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11393116 25 g 25g
11383116 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393116 Supplier Thermo Scientific Alfa Aesar Supplier No. L10988.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1455-21-6
Molecular Formula C9H20S
Molecular Weight (g/mol) 160.319
MDL Number MFCD00004913
InChI Key ZVEZMVFBMOOHAT-UHFFFAOYSA-N
Synonym 1-nonanethiol, n-nonyl mercaptan, nonyl mercaptan, nonylthiol, unii-39lg2etv2j, 39lg2etv2j, nonanethiol, non-thiol, n-nonylmercaptan, 1-nonylthiol
PubChem CID 15077
IUPAC Name nonane-1-thiol
SMILES CCCCCCCCCS

Spezifikation

Density 0.842
Boiling Point 219°C to 220°C
Flash Point 79°C (174°F)
Refractive Index 1.4548
Quantity 25 g
Beilstein 1733631
Sensitivity Air Sensitive
Formula Weight 160.32
Percent Purity 98%
Chemical Name or Material 1-Nonanethiol
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.