missing translation for 'onlineSavingsMsg'
Learn More

1-Nonanethiol, TRC

Product Code. 30480308
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30480308 500 mg 500mg
30467868 2.5 g 2.5g
30476825 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30480308 Supplier Toronto Research Chemicals Supplier No. TRCN649765500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1455-21-6
Molecular Formula C9 H20 S
Molecular Weight (g/mol) 160.32
Synonym 1-Mercaptononane, NSC 5563, Nonanethiol, Nonyl mercaptan, Nonylthiol, n-Nonanethiol, n-Nonyl mercaptan, n-Nonylthiol
IUPAC Name nonane-1-thiol
SMILES CCCCCCCCCS

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H20S/c1-2-3-4-5-6-7-8-9-10/h10H,2-9H2,1H3
Formula Weight 160.1286
Chemical Name or Material 1-Nonanethiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.