missing translation for 'onlineSavingsMsg'
Learn More

1-Nonanol, Extra Pure, SLR, Fisher Chemical

Product Code. 10000300 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10000300 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10000300 Supplier Fisher Chemical Supplier No. N/4020/08
  • ONLINE EXCLUSIVE PRICE
    187.00 €
    185.00 € / 500mL
    Save 2.00 €
    1% Off
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 143-08-8
Melting Point -5°C
Boiling Point 213°C
Packaging Amber glass bottle
Molecular Formula C9H20O
MDL Number 2990
Quantity 500 mL
Synonym 1-nonanol, nonanol, n-nonyl alcohol, nonyl alcohol, pelargonic alcohol, octyl carbinol, 1-hydroxynonane, nonalol, n-nonanol, n-nonan-1-ol
InChI Key ZWRUINPWMLAQRD-UHFFFAOYSA-N
SMILES CCCCCCCCCO
IUPAC Name nonan-1-ol
Molecular Weight (g/mol) 144.258
PubChem CID 8914
ChEBI CHEBI:35986
Formula Weight 144.26g/mol
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Nonanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.