missing translation for 'onlineSavingsMsg'
Learn More

1-Nonen-4-ol, 98%, Thermo Scientific Chemicals

Product Code. 11489786
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489786 25 g 25g
11479786 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489786 Supplier Thermo Scientific Alfa Aesar Supplier No. B20284.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35192-73-5
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00021949
InChI Key NDUPBTSXOSZHON-UHFFFAOYSA-N
Synonym 1-nonen-4-ol, 1-nonen-4-ol, 4s, nonen-4-ol, 4-hydroxy-1-nonene, allyl n-pentyl carbinol, acmc-20mpq3, acmc-1aftw
PubChem CID 141975
IUPAC Name non-1-en-4-ol
SMILES CCCCCC(CC=C)O

Specifications

Quantity 25 g
Formula Weight 142.24
Percent Purity 98%
Chemical Name or Material 1-Nonen-4-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.