missing translation for 'onlineSavingsMsg'
Learn More

1-O-Acetyl-2,3,5-tri-O-benzoyl-beta-D-ribofuranose, TRC

Product Code. 30328301
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
missing translation for 'unitSize':
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30328301 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30328301 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCA1893301G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-O-Acetyl-2,3,5-tri-O-benzoyl-beta-D-ribofuranose
CAS 6974-32-9
Molecular Formula C28 H24 O9
Quantity 1 g
Synonym β-D-Ribofuranose, 1-acetate 2,3,5-tribenzoate, Ribofuranose, 1-acetate 2,3,5-tribenzoate, β-D- (6CI,8CI), 1-0-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose, 1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose, 1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose, 1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribose, 1-O-Acetyltri-O-benzoyl-β-D-ribofuranose, 2,3,5-Tri-O-benzoyl-β-D-ribofuranosyl acetate, ABR, NSC 23349
InChI Formula InChI=1S/C28H24O9/c1-18(29)34-28-24(37-27(32)21-15-9-4-10-16-21)23(36-26(31)20-13-7-3-8-14-20)22(35-28)17-33-25(30)19-11-5-2-6-12-19/h2-16,22-24,28H,17H2,1H3/t22-,23-,24-,28-/m1/s1
SMILES CC(=O)O[C@@H]1O[C@H](COC(=O)c2ccccc2)[C@@H](OC(=O)c3ccccc3)[C@H]1OC(=O)c4ccccc4
IUPAC Name [(2R,3R,4R,5S)-5-acetyloxy-3,4-dibenzoyloxyoxolan-2-yl]methyl benzoate
Molecular Weight (g/mol) 504.48
Formula Weight 504.142
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.