missing translation for 'onlineSavingsMsg'
Learn More

1-O-n-Octyl-beta-D-glucopyranoside, 98%

Product Code. 10541794 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10541794 5 g Glass bottle 5g
10502251 250 mg Glass bottle 250mg
10442421 1 g Glass bottle 1g
10502174 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10541794 Supplier Thermo Scientific Acros Supplier No. 230340050
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 29836-26-8 | C14H28O6 | 292.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29836-26-8
Molecular Formula C14H28O6
Molecular Weight (g/mol) 292.37
MDL Number MFCD00063288
InChI Key HEGSGKPQLMEBJL-RKQHYHRCSA-N
Synonym octyl-beta-d-glucopyranoside, b-octylglucoside, octyl beta-d-glucopyranoside, octyl beta-d-glucoside, octyl glucoside, n-octyl-beta-d-glucoside, octyl b-d-glucopyranoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-octyloxy tetrahydro-2h-pyran-3,4,5-triol, unii-v109wut6rl, n-octyl glucoside
PubChem CID 62852
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-octoxyoxane-3,4,5-triol
SMILES CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 105°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 6857
Specific Rotation Condition − 32.00 (20.00°C c=5,water)
Specific Rotation -32
Solubility Information (10% in water) Clear to slightly hazy colorless to light yellow
Formula Weight 292.36
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 1-O-n-Octyl-β-D-glucopyranoside
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.