missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecanol, 95%

Product Code. 10534061 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
2.5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10534061 1 kg Plastic bottle 1kg
10667534 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10534061 Supplier Thermo Scientific Acros Supplier No. 129300010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 112-92-5 | C18H38O | 270.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-92-5
Molecular Formula C18H38O
Molecular Weight (g/mol) 270.5
MDL Number MFCD00002823
InChI Key GLDOVTGHNKAZLK-UHFFFAOYSA-N
Synonym stearyl alcohol, 1-octadecanol, octadecanol, octadecyl alcohol, n-octadecanol, 1-hydroxyoctadecane, n-1-octadecanol, stearol, n-octadecyl alcohol, stearic alcohol
PubChem CID 8221
ChEBI CHEBI:32154
IUPAC Name octadecan-1-ol
SMILES CCCCCCCCCCCCCCCCCCO

Specifications

Acidity 1mg KOH/g max.
CAS Min % 94.0
CAS Max % 100.0
Melting Point 55.0°C to 58.0°C
Color White
Density 185g/mL
Boiling Point 210.0°C (15.0 mmHg)
Flash Point 185°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Plastic bottle
Linear Formula CH3(CH2)17OH
Quantity 1 kg
Beilstein 01, 431
Merck Index 15, 8932
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and benzene
Formula Weight 270.5
Percent Purity 95%
Physical Form Flakes
Chemical Name or Material 1-Octadecanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.