missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecene, tech. 90%

Product Code. 11323136
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11323136 1000 mL 1 L
11313136 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11323136 Supplier Thermo Scientific Alfa Aesar Supplier No. L11004.AU
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 112-88-9 | C18H36 | 252.486 g/mol

1-Octadecene is an inexpensive solvent having high boiling point. Due to this property, it replaces the heat transfer fluids such as dowthem A or therminol 66 and used in the synthesis of high quality CdSe quantum dots (nano crystal made up of semiconductor materials). These quantum dots find application in transistors, diode lasers and solar cells. It is also used as a solvent for the synthesis of iron oxide nanospheres.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octadecene is an inexpensive solvent having high boiling point. Due to this property, it replaces the heat transfer fluids such as dowthem A or therminol 66 and used in the synthesis of high quality CdSe quantum dots (nano crystal made up of semiconductor materials). These quantum dots find application in transistors, diode lasers and solar cells. It is also used as a solvent for the synthesis of iron oxide nanospheres.

Solubility
Miscible with alcohol, acetone and ether. Immiscible with water.

Notes
Avoid reaction with strong Lewis or mineral acids. Free radical initiators should be avoided. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-88-9
Molecular Formula C18H36
Molecular Weight (g/mol) 252.486
MDL Number MFCD00009003
InChI Key CCCMONHAUSKTEQ-UHFFFAOYSA-N
Synonym 1-octadecene, octadecene, alpha-octadecene, .alpha.-octadecene, unii-h5zuq6v4ak, octadecene-1, h5zuq6v4ak, alkenes, c14-20 .alpha., 1-octadecene, analytical standard, linear octadecene
PubChem CID 8217
ChEBI CHEBI:30824
IUPAC Name octadec-1-ene
SMILES CCCCCCCCCCCCCCCCC=C

Specifications

Melting Point 15°C to 17°C
Density 0.789
Boiling Point 178°C to 179°C (15 mmHg)
Flash Point 154°C (309°F)
Refractive Index 1.444
Quantity 1000 mL
Beilstein 1761067
Solubility Information Miscible with alcohol,acetone and ether. Immiscible with water.
Formula Weight 252.48
Percent Purity 90%
Grade Technical
Chemical Name or Material 1-Octadecene
Show More Show Less
compliance-icons
Product Identifier
  • 1-Octadecene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: 1-Octadecene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.