missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecyne, 96%

Product Code. 15496097
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15496097 1 g 1g
15406107 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496097 Supplier Thermo Scientific Alfa Aesar Supplier No. H33288.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 629-89-0 | C18H34 | 250.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 629-89-0
Molecular Formula C18H34
Molecular Weight (g/mol) 250.47
MDL Number MFCD00015088
InChI Key IYDNQWWOZQLMRH-UHFFFAOYSA-N
Synonym 1-octadecyne, octadecyne, octadec-1-in, cetylacetylene, hexadecylacetylene, acmc-1b2rb, qspl 036
PubChem CID 69425
IUPAC Name octadec-1-yne
SMILES CCCCCCCCCCCCCCCCC#C

Spezifikation

Melting Point 21°C to 24°C
Density 0.803
Boiling Point 313°C
Quantity 1 g
Formula Weight 250.47
Percent Purity ≥97%
Chemical Name or Material 1-Octadecyne

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.