missing translation for 'onlineSavingsMsg'
Learn More

1-Octane Sulfonic Acid Sodium Salt, Ion Pair Chromatography, for Electrochemical Detection, Fisher Chemical

Product Code. 10588620 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic powder jar
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10588620 25 g Plastic powder jar 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10588620 Supplier Fisher Chemical Supplier No. O/0029/46
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 300°C
CAS 5324-84-5
Color White
pH 5.5 to 7.5
Physical Form Powder/Solid
Quantity 25 g
Packaging Plastic powder jar
Molecular Formula C8H17NaO3S
MDL Number MFCD00007544
Synonym sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
InChI Key HRQDCDQDOPSGBR-UHFFFAOYSA-M
SMILES [Na+].CCCCCCCCS([O-])(=O)=O
IUPAC Name sodium octane-1-sulfonate
Molecular Weight (g/mol) 216.27
PubChem CID 23669624
Formula Weight 216.28g/mol
Show More Show Less
compliance-icons
Product Identifier
  • 1-Octanesulfonic acid, sodium salt
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.