missing translation for 'onlineSavingsMsg'
Learn More

1-Octane Sulfonic Acid Sodium Salt, Ion Pair Chromatography, Fisher Chemical™

Product Code. 10151910 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Jar
Plastic powder jar
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10151910 25 g Plastic powder jar 25g
10450364 5 g Glass Jar 5g
10498790 100 g Plastic powder jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10151910 Supplier Fisher Chemical Supplier No. O/0028/46
  • ONLINE EXCLUSIVE PRICE
    219.45 €
    167.00 € / 25g
    Save 52.45 €
    23% Off
View all promotions
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

C8H17NaO3S, CAS Number-5324-84-5, 1-octanesulfonate, sodium, sodium octylsulfonate, ipc-alks-8, 1-octanesulfonic acid, sodium salt, 1-octanesulfonic acid sodium salt, octyl sodium sulfonate, sodium octanesulfonate, sodium octane-1-sulfonate, sodium 1-octanesulfonate, unii-j46ez5jyb3, 100g, White

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5324-84-5
Molecular Formula C8H17NaO3S
Molecular Weight (g/mol) 216.27
MDL Number MFCD00007544
InChI Key HRQDCDQDOPSGBR-UHFFFAOYSA-M
Synonym sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
PubChem CID 23669624
SMILES [Na+].CCCCCCCCS([O-])(=O)=O

Specifications

Melting Point 300°C
Color White
pH 5.5 to 7.5
Physical Form Powder/Solid
Quantity 25 g
IUPAC Name sodium;octane-1-sulfonate
Formula Weight 216.28g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • 1-Octanesulfonic acid, sodium salt
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.