missing translation for 'onlineSavingsMsg'
Learn More

1-Octane Sulfonic Acid Sodium Salt, Ion Pair Chromatography, Fisher Chemical™

Product Code. p-7073346 Shop All Fisher Chemical Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Jar
Plastic powder jar
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10450364

Brand: Fisher Chemical O/0028/44

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C8H17NaO3S, CAS Number-5324-84-5, 1-octanesulfonate, sodium, sodium octylsulfonate, ipc-alks-8, 1-octanesulfonic acid, sodium salt, 1-octanesulfonic acid sodium salt, octyl sodium sulfonate, sodium octanesulfonate, sodium octane-1-sulfonate, sodium 1-octanesulfonate, unii-j46ez5jyb3, 100g, White

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5324-84-5
Molecular Formula C8H17NaO3S
Molecular Weight (g/mol) 216.27
MDL Number MFCD00007544
InChI Key HRQDCDQDOPSGBR-UHFFFAOYSA-M
Synonym sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
PubChem CID 23669624
SMILES [Na+].CCCCCCCCS([O-])(=O)=O

Specifications

Melting Point 300°C
Color White
pH 5.5 to 7.5
Physical Form Powder/Solid
Quantity 5 g
IUPAC Name sodium octane-1-sulfonate
Formula Weight 216.28g/mol
Packaging Glass Jar
compliance-icons
Product Identifier
  • 1-Octanesulfonic acid, sodium salt
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.