missing translation for 'onlineSavingsMsg'
Learn More

1-Octanesulfonic acid, sodium salt, HPLC grade
GREENER_CHOICE

Product Code. 10386352 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10386352 5 g 5g
10529562 25 g 25g
10061822 100 g 100g
15264714 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10386352 Supplier Thermo Scientific Supplier No. 384770050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Octanesulfonic acid, sodium salt, HPLC grade, Quantity: 25g, White, Melting Point: >300.0 deg.C, Molecular Weight: 216.28, Percent Purity: >-98.0% | CAS: 5324-84-5 | C8H17NaO3S | 216.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 5324-84-5
Molecular Formula C8H17NaO3S
Molecular Weight (g/mol) 216.27
MDL Number MFCD00007544
InChI Key HRQDCDQDOPSGBR-UHFFFAOYSA-M
Synonym sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
PubChem CID 23669624
SMILES [Na+].CCCCCCCCS([O-])(=O)=O

Spécification

Absorbance (0.25M aqueous solution), (0.25 M aqueous solution) at 0nm, 0.04 max. at 260nm, 0.05 max. at 240nm, 0.05 max. at 250nm, 0.075 max. at 230nm, 0.1 max. at 220nm, 0.2 max. at 210nm, 0.3 max. at 200nm
Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula CH3(CH2)7SO3Na
Quantity 5 g
Solubility Information Solubility in water: freely soluble. Other solubilities: 4g/L in methanol
IUPAC Name sodium;octane-1-sulfonate
Formula Weight 216.28
Percent Purity ≥98.0%
Physical Form Crystalline Powder
Chemical Name or Material 1-Octanesulfonic acid, sodium salt
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • 1-Octanesulfonic acid, sodium salt
Mention d'avertissement
  • Warning
Catégorie de danger
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Informations supplémentaires
  • MIXTURE LIST-Contain: Sodium 1-octanesulfonate

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.