missing translation for 'onlineSavingsMsg'
Learn More

1-Octanesulfonic acid, sodium salt, HPLC grade

Product Code. 10529562 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10529562 25 g 25g
10386352 5 g 5g
10061822 100 g 100g
11306725 100 g 100g
15264714 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10529562 Supplier Thermo Scientific Supplier No. 384770250
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

1-Octanesulfonic acid, sodium salt, HPLC grade, Quantity: 25g, White, Melting Point: >300.0 deg.C, Molecular Weight: 216.28, Percent Purity: >-98.0% | CAS: 5324-84-5 | C8H17NaO3S | 216.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5324-84-5
Molecular Formula C8H17NaO3S
Molecular Weight (g/mol) 216.27
MDL Number MFCD00007544
InChI Key HRQDCDQDOPSGBR-UHFFFAOYSA-M
Synonym sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
PubChem CID 23669624
SMILES [Na+].CCCCCCCCS([O-])(=O)=O

Specifications

Absorbance (0.25M aqueous solution), (0.25 M aqueous solution) at 0nm, 0.04 max. at 260nm, 0.05 max. at 240nm, 0.05 max. at 250nm, 0.075 max. at 230nm, 0.1 max. at 220nm, 0.2 max. at 210nm, 0.3 max. at 200nm
Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula CH3(CH2)7SO3Na
Quantity 25 g
Solubility Information Solubility in water: freely soluble. Other solubilities: 4g/L in methanol
IUPAC Name sodium;octane-1-sulfonate
Formula Weight 216.28
Percent Purity ≥98.0%
Physical Form Crystalline Powder
Chemical Name or Material 1-Octanesulfonic acid, sodium salt
Show More Show Less
compliance-icons
Product Identifier
  • 1-Octanesulfonic acid, sodium salt
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.