missing translation for 'onlineSavingsMsg'
Learn More

1-Octanesulfonic Acid Sodium Salt, TRC

Product Code. 30409795
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30409795 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409795 Supplier Toronto Research Chemicals Supplier No. TRCO237575100G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Octanesulfonic Acid Sodium Salt
CAS 5324-84-5
Molecular Formula C8H17NaO3S
Quantity 100 g
Synonym Bio-Terge PAS 8, Bio-Terge PAS 8SF, NAS 8RF, NAS-FAL, Sodium 1-Octanesulfonate, Sodium Capryl Sulfonate, Sodium n-Octanesulfonate, Sodium n-Octylsulfonate, Sodium Octanesulfonate, Sodium Octylsulfonate, Witconate NAS-FAL
InChI Formula InChI=1S/C8H18O3S.Na/c1-2-3-4-5-6-7-8-12(9,10)11;/h2-8H2,1H3,(H,9,10,11);/q;+1/p-1
SMILES O=S(CCCCCCCC)(O[Na])=O
IUPAC Name sodium;octane-1-sulfonate
Molecular Weight (g/mol) 216.27
Formula Weight 216.0796
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.