missing translation for 'onlineSavingsMsg'
Learn More

1-Octanethiol, 97%

Product Code. 15562283 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2 kg
Unit Size:
2kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15562283 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 15562283 Supplier Thermo Scientific Acros Supplier No. 157480020
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 111-88-6 | C8H18S | 146.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 111-88-6
Melting Point -49.0°C
Density 0.8400g/mL
Boiling Point 197.0°C to 200.0°C
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Molecular Formula C8H18S
Linear Formula CH3(CH2)7SH
Refractive Index 1.4525 to 1.4545
MDL Number MFCD00004912
Quantity 2 kg
Specific Gravity 0.84
Synonym 1-octanethiol, 1-mercaptooctane, n-octyl mercaptan, octyl mercaptan, octanethiol, octylthiol, 1-octylthiol, n-octanethiol, octylmercaptan, n-octylthiol
InChI Key KZCOBXFFBQJQHH-UHFFFAOYSA-N
SMILES CCCCCCCCS
IUPAC Name octane-1-thiol
Molecular Weight (g/mol) 146.29
PubChem CID 8144
Formula Weight 146.29
Percent Purity 97%
Chemical Name or Material 1-Octanethiol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Octanethiol
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.